Learn More
ACROBiosystems Rhesus macaque CTGF / CCN2 Protein, His Tag
Catalog No. 502019693
Encompass_Preferred
Recombinant Protein;1MG;Rhesus macaque CTGF, His Tag (GTF-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ala 349 (Accession # H9FQD5-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.3 kDa. The protein migrates as 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rhesus macaque CTGF, His Tag (Cat. No. GTF-R52H4) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human CTGF Antibody, Human IgG1 with a linear range of 0.1-2 ng/mL (QC tested).
Catalog No. 50-201-9693 Supplier ACROBiosystems Supplier No. GTFR52H41MG
Recombinant Protein;1MG;Rhesus macaque CTGF, His Tag (GTF-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ala 349 (Accession # H9FQD5-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.3 kDa. The protein migrates as 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rhesus macaque CTGF, His Tag (Cat. No. GTF-R52H4) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human CTGF Antibody, Human IgG1 with a linear range of 0.1-2 ng/mL (QC tested).