Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) NSP7&NSP8 Protein, His Tag
Catalog No. 502019697
Encompass_Preferred
Recombinant Protein;1MG;SARS-CoV-2 NSP7&NSP8, His Tag (NS8-C5125) is expressed from E.coli cells. It contains AA Ser 1 - Gln 83 (NSP7) & Ala 1 - Gln 198 (NSP8) (Accession # YP_009725303.1 (NSP7) & YP_009725304.1 (NSP8)).;This protein carries a polyhistidine tag.The protein has a calculated MW of 32.1 kDa. The protein migrates as 28-32 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-201-9697 Supplier ACROBiosystems Supplier No. NS8C51251MG
Recombinant Protein;1MG;SARS-CoV-2 NSP7&NSP8, His Tag (NS8-C5125) is expressed from E.coli cells. It contains AA Ser 1 - Gln 83 (NSP7) & Ala 1 - Gln 198 (NSP8) (Accession # YP_009725303.1 (NSP7) & YP_009725304.1 (NSP8)).;This protein carries a polyhistidine tag.The protein has a calculated MW of 32.1 kDa. The protein migrates as 28-32 kDa under reducing (R) condition (SDS-PAGE).