Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) NSP16&NSP10 Heterodimer Protein, His Tag&Twin Strep Tag
Catalog No. 502019732
Encompass_Preferred
Recombinant Protein;1MG;SARS-CoV-2 NSP16&NSP10 Heterodimer Protein, His Tag&Twin Strep Tag (NS0-C51W3) is expressed from E.coli cells. It contains AA Ser 1 - Asn 298 (NSP16) & Ala 1 - Gln 139 (NSP10) (Accession # YP_009725311.1 (NSP16) & YP_009725306.1 (NSP10)).;SARS-CoV-2 NSP16&NSP10 Heterodimer Protein, His Tag&Twin Strep Tag is produced by co-expression of NSP16 and NSP10, has a calculated MW of 35.3 kDa (NSP16) and 18.3 kDa (NSP10). Subunit NSP16 is fused with a polyhistidine tag at the N-terminus and subunit NSP10 is fused with a Twin Strep tag at the N-terminus. The reducing (R) heterodimer protein migrates as 18-19 kDa and 35 kDa.
Catalog No. 50-201-9732 Supplier ACROBiosystems Supplier No. NS0C51W31MG
Recombinant Protein;1MG;SARS-CoV-2 NSP16&NSP10 Heterodimer Protein, His Tag&Twin Strep Tag (NS0-C51W3) is expressed from E.coli cells. It contains AA Ser 1 - Asn 298 (NSP16) & Ala 1 - Gln 139 (NSP10) (Accession # YP_009725311.1 (NSP16) & YP_009725306.1 (NSP10)).;SARS-CoV-2 NSP16&NSP10 Heterodimer Protein, His Tag&Twin Strep Tag is produced by co-expression of NSP16 and NSP10, has a calculated MW of 35.3 kDa (NSP16) and 18.3 kDa (NSP10). Subunit NSP16 is fused with a polyhistidine tag at the N-terminus and subunit NSP10 is fused with a Twin Strep tag at the N-terminus. The reducing (R) heterodimer protein migrates as 18-19 kDa and 35 kDa.