Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) NSP7 Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems NS7C51H61MG
Encompass_Preferred
Recombinant Protein;1MG;SARS-CoV-2 NSP7, His Tag (NS7-C51H6) is expressed from E.coli cells. It contains AA Ser 1 - Gln 83 (Accession # YP_009725303.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 11.3 kDa. The protein migrates as 12 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-201-9736