Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) NSP8 Protein, His Tag
Supplier:  ACROBiosystems NS8C51491MG
Encompass_Preferred
Recombinant Protein;1MG;SARS-CoV-2 NSP8, His Tag (NS8-C5149) is expressed from E.coli cells. It contains AA Ala 1 - Gln 198 (Accession # YP_009725304.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 23.9 kDa. The protein migrates as 22-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9738