Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) NSP1 Protein, His Tag
Catalog No. 502019734
Encompass_Preferred
Recombinant Protein;1MG;SARS-CoV-2 NSP1, His Tag (NS1-C51H7) is expressed from E.coli cells. It contains AA Met 1 - Gly 180 (Accession # YP_009725297.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.7 kDa. The protein migrates as 22-25 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-201-9734 Supplier ACROBiosystems Supplier No. NS1C51H71MG
Recombinant Protein;1MG;SARS-CoV-2 NSP1, His Tag (NS1-C51H7) is expressed from E.coli cells. It contains AA Met 1 - Gly 180 (Accession # YP_009725297.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.7 kDa. The protein migrates as 22-25 kDa under reducing (R) condition (SDS-PAGE).