Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S2 protein, His Tag
Supplier:  ACROBiosystems S2NC52H21MG
Encompass_Preferred
Recombinant Protein;1MG;SARS-CoV-2 S2 protein, His Tag (S2N-C52H2) is expressed from human 293 cells (HEK293). It contains AA Ser 686 - Pro1213 (Accession # QHD43416.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.0 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9511