Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems SARS-CoV-2 Papain-Like Protease, His Tag, 1 mg

Catalog No. 502019752
Encompass_Preferred

This SARS-CoV-2 papain-like protease (PL-PRO) is an active enzyme expressed in E. coli that plays a critical role in the processing of viral polyproteins. By cleaving at least 11 sites on the large polyprotein 1ab, this enzyme is essential for viral replication. This product contains a polyhistidine tag at the N-terminus and is provided as a 0.2 μm filtered liquid solution.

  • Purity >95% as determined by SDS-PAGE
  • Calculated molecular weight of 37.6 kDa (migrates at 35-37 kDa under reducing conditions)
  • Specific activity ≥2500 pmol/min/mg as measured by substrate conversion
  • Endotoxin level <1.0 EU/μg via LAL method
  • Supplied in 50 mM HEPES, 300 mM NaCl, pH 7.5, and trehalose with glycerol

Catalog No. 50-201-9752 Supplier ACROBiosystems Supplier No. PAEC51481MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This SARS-CoV-2 papain-like protease (PL-PRO) is an active enzyme expressed in E. coli that plays a critical role in the processing of viral polyproteins. By cleaving at least 11 sites on the large polyprotein 1ab, this enzyme is essential for viral replication. This product contains a polyhistidine tag at the N-terminus and is provided as a 0.2 μm filtered liquid solution.

  • Purity >95% as determined by SDS-PAGE
  • Calculated molecular weight of 37.6 kDa (migrates at 35-37 kDa under reducing conditions)
  • Specific activity ≥2500 pmol/min/mg as measured by substrate conversion
  • Endotoxin level <1.0 EU/μg via LAL method
  • Supplied in 50 mM HEPES, 300 mM NaCl, pH 7.5, and trehalose with glycerol

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.