Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) Nucleocapsid protein, His Tag
Supplier:  ACROBiosystems NUNC51H21MG
Encompass_Preferred
Recombinant Protein;1MG;SARS-CoV-2 Nucleocapsid protein, His Tag (NUN-C51H2) is expressed from E.coli cells. It contains AA Met 1 - Ala 419 (Accession # QHO62115.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 51.0 kDa. The protein migrates as 53-55 kDa under reducing (R) condition (SDS-PAGE).;Immobilized SARS-CoV-2 Nucleocapsid protein, His Tag (Cat. No. NUN-C51H2) at 2 μg/mL (100 μL/well) can bind 2019-nCoV NP Antibody, Rabbit MAb (CLN27) with a linear range of 0.02-0.6 ng/mL (QC tested).
Catalog No. 50-201-9669