Learn More
ACROBiosystems Biotinylated Human Transferrin R / CD71 Protein, His,Avitag™
Supplier:  ACROBiosystems TFRH82E5200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human Transferrin R, His,Avitag™ (TFR-H82E5) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession # P02786-1).;This protein carries an Avi tag at the N-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 78.7 kDa.
Catalog No. 50-201-9403