Learn More
ACROBiosystems Biotinylated Human TGF-beta RII / TGFBR2 Protein, His,Avitag™
Catalog No. 502019561
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human TGF-beta RII, His,Avitag (TG2-H82E4) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Asp 159 (Accession # P37173-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 19.1 kDa. The protein migrates as 28-40 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Human Latent TGFB1, His Tag (Cat. No. TG1-H524x) at 1 μg/mL (100 μL/well) can bind Biotinylated Human TGF-beta RII, His,Avitag (Cat. No. TG2-H82E4) with a linear range of 5-78 ng/mL (QC tested).
Catalog No. 50-201-9561 Supplier ACROBiosystems Supplier No. TG2H82E4200UG
Recombinant Protein;200UG;Biotinylated Human TGF-beta RII, His,Avitag (TG2-H82E4) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Asp 159 (Accession # P37173-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 19.1 kDa. The protein migrates as 28-40 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Human Latent TGFB1, His Tag (Cat. No. TG1-H524x) at 1 μg/mL (100 μL/well) can bind Biotinylated Human TGF-beta RII, His,Avitag (Cat. No. TG2-H82E4) with a linear range of 5-78 ng/mL (QC tested).