Learn More
ACROBiosystems Biotinylated Human TGF-beta RII / TGFBR2 Protein, Fc,Avitag™ (MALS verified)
Supplier:  ACROBiosystems TG2H82F6200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human TGFBR2, Fc,Avitag (TG2-H82F6) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Asp 159 (Accession # P37173-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 43.7 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Latent TGFB1, His Tag (Cat. No. TG1-H524x) at 2 μg/mL (100 μL/well) can bind Biotinylated Human TGFBR2, Fc,Avitag (Cat. No. TG2-H82F6) with a linear range of 2-16 ng/mL (QC tested).
Catalog No. 50-201-9614