Learn More
ACROBiosystems Biotinylated Human CD59 Protein, His,Avitag™
Catalog No. 502019628
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CD59, His,Avitag (CD9-H82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Asn 102 (Accession # P13987-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 12.5 kDa. The protein migrates as 15-20 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9628 Supplier ACROBiosystems Supplier No. CD9H82E3200UG
Recombinant Protein;200UG;Biotinylated Human CD59, His,Avitag (CD9-H82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Asn 102 (Accession # P13987-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 12.5 kDa. The protein migrates as 15-20 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.