Learn More
ACROBiosystems Biotinylated Human BAFFR / TNFRSF13C Protein, His,Avitag™
Catalog No. 502019421
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Mouse BAFFR, His,Avitag™ (BAR-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Ala 71 (Accession # Q96RJ3-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 10.1 kDa.
Catalog No. 50-201-9421 Supplier ACROBiosystems Supplier No. BARH82E3200UG
Recombinant Protein;200UG;Biotinylated Mouse BAFFR, His,Avitag™ (BAR-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Ala 71 (Accession # Q96RJ3-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 10.1 kDa.