Learn More
ACROBiosystems Biotinylated Human GUCY2C / Guanylyl cyclase C Protein, His,Avitag™
Catalog No. 502019648
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human GUCY2C, His,Avitag (GUC-H82E9) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Gln 430 (Accession # P25092-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 49.6 kDa. The protein migrates as 65-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human GUCY2C, His,Avitag (Cat. No. GUC-H82E9) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Monoclonal Anti-Human GUCY2C Antibody, Human IgG1 with a linear range of 0.2-2 ng/mL (QC tested).;Immobilized Monoclonal Anti-Human GUCY2C Antibody, Human IgG1 at 1 μg/mL (100 μL/well) can bind Biotinylated Human GUCY2C, His,Avitag (Cat. No. GUC-H82E9) with a linear range of 0.2-6 ng/mL (Routinely tested).
Catalog No. 50-201-9648 Supplier ACROBiosystems Supplier No. GUCH82E9200UG
Recombinant Protein;200UG;Biotinylated Human GUCY2C, His,Avitag (GUC-H82E9) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Gln 430 (Accession # P25092-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 49.6 kDa. The protein migrates as 65-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human GUCY2C, His,Avitag (Cat. No. GUC-H82E9) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Monoclonal Anti-Human GUCY2C Antibody, Human IgG1 with a linear range of 0.2-2 ng/mL (QC tested).;Immobilized Monoclonal Anti-Human GUCY2C Antibody, Human IgG1 at 1 μg/mL (100 μL/well) can bind Biotinylated Human GUCY2C, His,Avitag (Cat. No. GUC-H82E9) with a linear range of 0.2-6 ng/mL (Routinely tested).