Learn More
ACROBiosystems Biotinylated Human CD8 alpha / CD8A Protein, His,Avitag™
Catalog No. 502019724
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CD8 alpha, His,Avitag (CDA-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Asp 182 (Accession # P01732-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 21.2 kDa. The protein migrates as 25-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human CD8 alpha, His,Avitag (Cat. No. CDA-H82E3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate. can bind Anti-CD8 alpha Antibody, Mouse IgG2a (clone: OKT8) with a linear range of 0.2-2 ng/mL (QC tested).;Immobilized Anti-CD8 alpha Antibody, Mouse IgG2a (clone: OKT8) at 1 μg/mL (100 μL/well) can bind Biotinylated Human CD8 alpha, His,Avitag (Cat. No. CDA-H82E3) with a linear range of 1-39 ng/mL (Routinely tested).
Catalog No. 50-201-9724 Supplier ACROBiosystems Supplier No. CDAH82E3200UG
Recombinant Protein;200UG;Biotinylated Human CD8 alpha, His,Avitag (CDA-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Asp 182 (Accession # P01732-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 21.2 kDa. The protein migrates as 25-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human CD8 alpha, His,Avitag (Cat. No. CDA-H82E3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate. can bind Anti-CD8 alpha Antibody, Mouse IgG2a (clone: OKT8) with a linear range of 0.2-2 ng/mL (QC tested).;Immobilized Anti-CD8 alpha Antibody, Mouse IgG2a (clone: OKT8) at 1 μg/mL (100 μL/well) can bind Biotinylated Human CD8 alpha, His,Avitag (Cat. No. CDA-H82E3) with a linear range of 1-39 ng/mL (Routinely tested).