Learn More
ACROBiosystems Biotinylated Human CD36 / SR-B3 Protein, His,Avitag™
Catalog No. 502019766
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CD36, His,Avitag (CD6-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Asn 439 (Accession # NP_001001547.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 50.2 kDa.
Catalog No. 50-201-9766 Supplier ACROBiosystems Supplier No. CD6H82E9200UG
Recombinant Protein;200UG;Biotinylated Human CD36, His,Avitag (CD6-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Asn 439 (Accession # NP_001001547.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 50.2 kDa.