Learn More
ACROBiosystems Biotinylated Mouse CD28 Protein, His,Avitag™
Catalog No. 502019764
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Mouse CD28, His,Avitag (CD8-M82E3) is expressed from human 293 cells (HEK293). It contains AA Asn 20 - Lys 149 (Accession # P31041-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 18.6 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Mouse CD28, His,Avitag (Cat. No. CD8-M82E3) at 1 μg/mL (100 μL/well) on Streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Mouse B7-1, Fc Tag (Cat. No. CD0-M5259) with a linear range of 0.6-10 ng/mL (QC tested).;Immobilized Biotinylated Mouse CD28, His,Avitag (Cat. No. CD8-M82E3) at 1 μg/mL (100 μL/well) on Streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Human B7-1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. B71-H52A4) with a linear range of 0.2-5 ng/mL (Routinely tested).
Catalog No. 50-201-9764 Supplier ACROBiosystems Supplier No. CD8M82E3200UG
Recombinant Protein;200UG;Biotinylated Mouse CD28, His,Avitag (CD8-M82E3) is expressed from human 293 cells (HEK293). It contains AA Asn 20 - Lys 149 (Accession # P31041-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 18.6 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Mouse CD28, His,Avitag (Cat. No. CD8-M82E3) at 1 μg/mL (100 μL/well) on Streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Mouse B7-1, Fc Tag (Cat. No. CD0-M5259) with a linear range of 0.6-10 ng/mL (QC tested).;Immobilized Biotinylated Mouse CD28, His,Avitag (Cat. No. CD8-M82E3) at 1 μg/mL (100 μL/well) on Streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Human B7-1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. B71-H52A4) with a linear range of 0.2-5 ng/mL (Routinely tested).