Learn More
ACROBiosystems Biotinylated Human CD39 / ENTPD1 Protein, His,Avitag™ (MALS verified)
Catalog No. 502019820
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CD39, His,Avitag (MALS verified) (CD9-H82E5) is expressed from human 293 cells (HEK293). It contains AA Thr 38 - Val 478 (Accession # P49961-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 54.0 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9820 Supplier ACROBiosystems Supplier No. CD9H82E5200UG
Recombinant Protein;200UG;Biotinylated Human CD39, His,Avitag (MALS verified) (CD9-H82E5) is expressed from human 293 cells (HEK293). It contains AA Thr 38 - Val 478 (Accession # P49961-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 54.0 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.