Learn More
ACROBiosystems Biotinylated Human CD38 Protein, Avitag™,His Tag
Supplier:  ACROBiosystems CD8H82E7200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CD38, Avitag,His Tag (CD8-H82E7) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Accession # NP_001766.2).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 32.5 kDa. The protein migrates as 40-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-CD38 MAb, Human IgG1 at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD38, Avitag,His Tag (Cat. No. CD8-H82E7) with a linear range of 0.08-1.28 μg/mL (QC tested).
Catalog No. 50-201-9849