Learn More
ACROBiosystems Biotinylated Human FAP Protein, His,Avitag™
Supplier:  ACROBiosystems FAPH82Q6200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human FAP Protein, His,Avitag™ (FAP-H82Q6) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Asp 760 (Accession # Q12884-1).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 88.6 kDa.
Catalog No. 50-201-9477