Learn More
ACROBiosystems Biotinylated Human CD229 / SLAMF3 Protein, His,Avitag™
Supplier:  ACROBiosystems CD9H82E6200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CD229, His,Avitag™ (CD9-H82E6) is expressed from human 293 cells (HEK293). It contains AA Lys 48 - Lys 454 (Accession # Q9HBG7-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 48.4 kDa. The protein migrates as 65-70 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.
Catalog No. 50-201-9494