Learn More
ACROBiosystems Biotinylated Cynomolgus / Rhesus macaque BCMA / TNFRSF17 Protein, His,Avitag™
Catalog No. 502019540
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Cynomolgus / Rhesus macaque BCMA, His,Avitag (BCA-C82E5) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession # G7Q0I4-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 9.6 kDa. The protein migrates as 15-18 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human APRIL, His,Flag Tag, active trimer (Cat. No. APL-H52D1) at 2 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus / Rhesus macaque BCMA, His,Avitag (Cat. No. BCA-C82E5) with a linear range of 0.2-1 ng/mL (QC tested).;Immobilized Biotinylated Cynomolgus / Rhesus macaque BCMA, His,Avitag (Cat. No. BCA-C82E5) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) with a linear range of 0.2-5 ng/mL (Routinely tested).
Catalog No. 50-201-9540 Supplier ACROBiosystems Supplier No. BCAC82E5200UG
Recombinant Protein;200UG;Biotinylated Cynomolgus / Rhesus macaque BCMA, His,Avitag (BCA-C82E5) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession # G7Q0I4-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 9.6 kDa. The protein migrates as 15-18 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human APRIL, His,Flag Tag, active trimer (Cat. No. APL-H52D1) at 2 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus / Rhesus macaque BCMA, His,Avitag (Cat. No. BCA-C82E5) with a linear range of 0.2-1 ng/mL (QC tested).;Immobilized Biotinylated Cynomolgus / Rhesus macaque BCMA, His,Avitag (Cat. No. BCA-C82E5) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) with a linear range of 0.2-5 ng/mL (Routinely tested).