Learn More
ACROBiosystems FITC-Labeled Human CLEC12A / MICL / CLL-1 Protein, His Tag
Supplier:  ACROBiosystems CLAHF247200UG
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human CLEC12A, His Tag (CLA-HF247) is expressed from human 293 cells (HEK293). It contains AA His 65 - Ala 265 (Accession # Q5QGZ9-2). It is the FITC labeled form of Human CLEC12A, His Tag (CLA-H5245).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 25.6 kDa. The protein migrates as 35-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;2e5 of Anti-CLEC12A CAR-293 cells were stained with 100 μL of 10 μg/mL of FITC-Labeled Human CLEC12A, His Tag (Cat. No. CLA-HF247) and negative control protein respectively. FITC signal was used to evaluate the binding activity (QC tested).
Catalog No. 50-201-9689