Learn More
ACROBiosystems FITC-Labeled Human BCMA / TNFRSF17 Protein, Fc Tag
Catalog No. 502019838
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human BCMA, Fc Tag (Cat. No. BCA-HF254) is expressed from human HEK293 cells. It contains AA Met 1 - Ala 54 (Accession # Q02223-1). It is the FITC labeled form of Human BCMA, Fc Tag (Cat. No. BC7-H5254).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 32.0 kDa. As a result of glycosylation, the protein migrates as 35-44 kDa under reducing (R) condition, and 60-75 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human BAFF, Fc Tag (Cat. No. BAF-H4268) at 10 μg/mL (100 μL/well) can bind FITC-Labeled Human BCMA, Fc Tag (Cat. No. BCA-HF254) with a linear range of 0.04-1.25 μg/mL (Ex.488 nm/Em.535 nm) (QC tested).;293 cells were transfected with anti-BCMA-scFv and RFP tag. 2e5 of the cells were stained with B. FITC-labeled human BCMA / TNFRSF17 protein, Fc tag (Cat. No. BCA-HF254, 3 μg/ml) and C. FITC-labeled protein control. A. Non-transfected 293 cells and C. FITC-labeled protein control wer
Catalog No. 50-201-9838 Supplier ACROBiosystems Supplier No. BCAHF254200UG
Recombinant Protein;200UG;FITC-Labeled Human BCMA, Fc Tag (Cat. No. BCA-HF254) is expressed from human HEK293 cells. It contains AA Met 1 - Ala 54 (Accession # Q02223-1). It is the FITC labeled form of Human BCMA, Fc Tag (Cat. No. BC7-H5254).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 32.0 kDa. As a result of glycosylation, the protein migrates as 35-44 kDa under reducing (R) condition, and 60-75 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human BAFF, Fc Tag (Cat. No. BAF-H4268) at 10 μg/mL (100 μL/well) can bind FITC-Labeled Human BCMA, Fc Tag (Cat. No. BCA-HF254) with a linear range of 0.04-1.25 μg/mL (Ex.488 nm/Em.535 nm) (QC tested).;293 cells were transfected with anti-BCMA-scFv and RFP tag. 2e5 of the cells were stained with B. FITC-labeled human BCMA / TNFRSF17 protein, Fc tag (Cat. No. BCA-HF254, 3 μg/ml) and C. FITC-labeled protein control. A. Non-transfected 293 cells and C. FITC-labeled protein control wer