Learn More
ACROBiosystems FITC-Labeled Human Siglec-3 / CD33 Protein, Fc Tag
Catalog No. 502019663
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human Siglec-3, Fc Tag (CD3-HF256) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1). It is the FITC labeled form of Human Siglec-3, Fc Tag (CD3-H5257).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.9 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-3 Antibody at 1 μg/mL (100 μL/well) can bind FITC-Labeled Human Siglec-3, Fc Tag (Cat. No. CD3-HF256) with a linear range of 0.313-5 μg/mL (QC tested).
Catalog No. 50-201-9663 Supplier ACROBiosystems Supplier No. CD3HF256200UG
Recombinant Protein;200UG;FITC-Labeled Human Siglec-3, Fc Tag (CD3-HF256) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1). It is the FITC labeled form of Human Siglec-3, Fc Tag (CD3-H5257).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.9 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-3 Antibody at 1 μg/mL (100 μL/well) can bind FITC-Labeled Human Siglec-3, Fc Tag (Cat. No. CD3-HF256) with a linear range of 0.313-5 μg/mL (QC tested).