Learn More
ACROBiosystems FITC-Labeled Human FOLR1 Protein, His Tag
Supplier:  ACROBiosystems FO1HF2H8200UG
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human FOLR1, His Tag (FO1-HF2H8) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # P15328-1). It is the FITC labeled form of Human FOLR1, His Tag (FO1-H52H1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa. The protein migrates as 35-43 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Folic acid-BSA conjugate at 5 μg/mL (100 μL/well) can bind FITC-Labeled Human FOLR1, His Tag (Cat. No. FO1-HF2H8) with a linear range of 0.078-1.25 μg/mL (QC tested).
Catalog No. 50-201-9683