Learn More
ACROBiosystems FITC-Labeled Human CD30 / TNFRSF8 Protein, Fc Tag
Catalog No. 502019432
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human CD30, His Tag (Cat. No. CD0-HF250) is expressed from human HEK293 cells. It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2). It is the FITC labeled form of Human CD30, His Tag (Cat. No.CD0-H5250).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64.6 kDa. The protein migrates as 70-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Monoclonal Anti-Human CD30 Human Antibody, Human IgG1 at 1 μg/mL (100 μL/well) can bind FITC-Labeled Human CD30, Fc Tag (Cat. No. CD0-HF250) with a linear range of 0.08-1.3 μg/mL (QC tested).;Immobilized Human CD30 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-H525b) at 2 μg/mL (100 μL/well) can bind FITC-Labeled Human CD30, Fc Tag (Cat. No. CD0-HF250) with a linear range of 0.2-1.3 μg/mL (Routinely tested).
Catalog No. 50-201-9432 Supplier ACROBiosystems Supplier No. CD0HF250200UG
Recombinant Protein;200UG;FITC-Labeled Human CD30, His Tag (Cat. No. CD0-HF250) is expressed from human HEK293 cells. It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2). It is the FITC labeled form of Human CD30, His Tag (Cat. No.CD0-H5250).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64.6 kDa. The protein migrates as 70-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Monoclonal Anti-Human CD30 Human Antibody, Human IgG1 at 1 μg/mL (100 μL/well) can bind FITC-Labeled Human CD30, Fc Tag (Cat. No. CD0-HF250) with a linear range of 0.08-1.3 μg/mL (QC tested).;Immobilized Human CD30 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-H525b) at 2 μg/mL (100 μL/well) can bind FITC-Labeled Human CD30, Fc Tag (Cat. No. CD0-HF250) with a linear range of 0.2-1.3 μg/mL (Routinely tested).