Learn More
ACROBiosystems Human TROP-2 / TACSTD2 Protein, Fc Tag
Catalog No. 502019803
Encompass_Preferred
Recombinant Protein;200UG;Human TROP-2, Fc Tag (TR2-H5253) is expressed from human 293 cells (HEK293). It contains AA His 27 - Thr 274 (Accession # P09758-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 54.3 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9803 Supplier ACROBiosystems Supplier No. TR2H5253200UG
Recombinant Protein;200UG;Human TROP-2, Fc Tag (TR2-H5253) is expressed from human 293 cells (HEK293). It contains AA His 27 - Thr 274 (Accession # P09758-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 54.3 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.