Learn More
ACROBiosystems Mouse TROP-2 / TACSTD2 Protein, His Tag
Catalog No. 502019558
Encompass_Preferred
Recombinant Protein;200UG;Mouse TROP-2, His Tag (TR2-M52H6) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Gly 270 (Accession # Q8BGV3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.9 kDa. The protein migrates as 38-48 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.
Catalog No. 50-201-9558 Supplier ACROBiosystems Supplier No. TR2M52H6200UG
Recombinant Protein;200UG;Mouse TROP-2, His Tag (TR2-M52H6) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Gly 270 (Accession # Q8BGV3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.9 kDa. The protein migrates as 38-48 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.