Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) Nucleocapsid protein, His Tag
Catalog No. 502019528
Encompass_Preferred
Recombinant Protein;200UG;SARS-CoV-2 Nucleocapsid protein, His Tag (NUN-C5221) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.3 kDa.
Catalog No. 50-201-9528 Supplier ACROBiosystems Supplier No. NUNC5221200UG
Recombinant Protein;200UG;SARS-CoV-2 Nucleocapsid protein, His Tag (NUN-C5221) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.3 kDa.