Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S protein RBD, His Tag
Supplier: ACROBiosystems SPDC52H1200UG
Recombinant Protein;200UG;SARS-CoV-2 S protein RBD, His Tag (SPD-C52H1) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa. The protein migrates as 32-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 S protein RBD, His Tag (Cat. No. SPD-C52H1) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Neutralizing Antibody, Human IgG1 (Cat. No. SAD-S35) with a linear range of 0.1-3 ng/mL (QC tested).