Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S1 protein, His Tag
Supplier:  ACROBiosystems S1NC52H2200UG
Encompass_Preferred
Recombinant Protein;200UG;SARS-CoV-2 S1 protein, His Tag (S1N-C52H2) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 76.9 kDa. The protein migrates as 110-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 S1 protein, His Tag (Cat. No. S1N-C52H2) at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-SARS-CoV-S protein RBD Antibody, Human IgG1 with a linear range of 0.2-6 ng/mL (QC tested).
Catalog No. 50-201-9675