Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S protein RBD, His Tag
Catalog No. 502019420
Encompass_Preferred
Recombinant Protein;200UG;SARS-CoV-2 S protein RBD, His Tag (SPD-C52H2) is expressed from human 293 cells (HEK293).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.7 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized SARS-CoV-2 S protein RBD, His Tag (Cat. No. SPD-C52H2) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 RBD Neutralizing Antibody, Human IgG1 (Cat. No. SAD-S35) with a linear range of 0.2-3 ng/mL (QC tested).
Catalog No. 50-201-9420 Supplier ACROBiosystems Supplier No. SPDC52H2200UG
Recombinant Protein;200UG;SARS-CoV-2 S protein RBD, His Tag (SPD-C52H2) is expressed from human 293 cells (HEK293).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.7 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized SARS-CoV-2 S protein RBD, His Tag (Cat. No. SPD-C52H2) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 RBD Neutralizing Antibody, Human IgG1 (Cat. No. SAD-S35) with a linear range of 0.2-3 ng/mL (QC tested).