Learn More
ACROBiosystems Human IL-17F / IL-24 Protein, His Tag
Supplier:  ACROBiosystems ILFH524420UG
Encompass_Preferred
Recombinant Protein;20UG;Human IL-17F, His Tag (ILF-H5244) is expressed from human 293 cells (HEK293). It contains AA Arg 31 - Gln 163 (Accession # Q96PD4-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 22-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-17F, His Tag (Cat. No. ILF-H5244) at 5 μg/mL (100 μL/well) can bind Human IL-17 RA, Fc Tag (Cat. No. ILA-H5257) with a linear range of 0.02-0.625 μg/mL (QC tested).;Immobilized Human IL-17F, His Tag (Cat. No. ILF-H5244) at 5 μg/mL (100 μL/well) can bind Human IL-17 RC, Fc Tag (Cat. No. ILC-H5255) with a linear range of 0.039-1.25 μg/mL (Routinely tested).
Catalog No. 50-201-9719