Learn More
ACROBiosystems Human CD20 / MS4A1 Full Length Protein, His Tag (Nanodisc) (HEK293)
Catalog No. 502019819
Encompass_Preferred
Recombinant Protein;20UG;Human CD20 Full Length, His Tag (CD0-H52H1) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Pro 297 (Accession # P11836-1).;Nanodiscs are a new class of model membranes that are being used to solubilize and study a range of integral membrane proteins (mp,full length CD20) and membrane-associated proteins. The Nanodisc bilayer is bounded by a membrane scaffold protein (MSP1D1) coat that confers enhanced stability and a narrow particle size distribution.
The CD20 nanodisc assembles from a mixture of full length CD20 protein in detergent, phospholipid micelles and membrane scaffold protein(MSP1D1) upon removal of the detergent.
The CD20 carries a polyhistidine tag at the C-terminus with calculated MW of 35.2 kDa and migrates as 40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation. The membrane scaffold protein(MSP1D1)has calculated MW of 24.7 kDa, and it migrates as 25 kDa under reducing (R) condition (SDS-PAGE) .;Immob
Catalog No. 50-201-9819 Supplier ACROBiosystems Supplier No. CD0H52H120UG
Recombinant Protein;20UG;Human CD20 Full Length, His Tag (CD0-H52H1) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Pro 297 (Accession # P11836-1).;Nanodiscs are a new class of model membranes that are being used to solubilize and study a range of integral membrane proteins (mp,full length CD20) and membrane-associated proteins. The Nanodisc bilayer is bounded by a membrane scaffold protein (MSP1D1) coat that confers enhanced stability and a narrow particle size distribution.
The CD20 nanodisc assembles from a mixture of full length CD20 protein in detergent, phospholipid micelles and membrane scaffold protein(MSP1D1) upon removal of the detergent.
The CD20 carries a polyhistidine tag at the C-terminus with calculated MW of 35.2 kDa and migrates as 40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation. The membrane scaffold protein(MSP1D1)has calculated MW of 24.7 kDa, and it migrates as 25 kDa under reducing (R) condition (SDS-PAGE) .;Immob