Learn More
ACROBiosystems Human IL-10 Protein, His Tag
Supplier:  ACROBiosystems IL0H424820UG
Encompass_Preferred
Recombinant Protein;20UG;Human IL-10, His Tag (IL0-H4248) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Asn 178 (Accession # NP_000563.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 19.5 kDa. The protein migrates as 19 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9869