Learn More
ACROBiosystems Biotinylated Human IL-5 Protein, His,Avitag™
Catalog No. 502019571
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human IL-5, His,Avitag (IL5-H82Q5) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Ser 134 (Accession # P05113-1).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 16.7 kDa. The protein migrates as 20-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.; Immobilized Biotinylated Human IL-5, His,Avitag (Cat. No. IL5-H82Q5) at 1 μg/mL (100 μL/well) can bind Human IL-5 R alpha, Fc Tag (Cat. No. ILA-H5259) with a linear range of 5-78 ng/mL (QC tested).
Catalog No. 50-201-9571 Supplier ACROBiosystems Supplier No. IL5H82Q525UG
Recombinant Protein;25UG;Biotinylated Human IL-5, His,Avitag (IL5-H82Q5) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Ser 134 (Accession # P05113-1).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 16.7 kDa. The protein migrates as 20-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.; Immobilized Biotinylated Human IL-5, His,Avitag (Cat. No. IL5-H82Q5) at 1 μg/mL (100 μL/well) can bind Human IL-5 R alpha, Fc Tag (Cat. No. ILA-H5259) with a linear range of 5-78 ng/mL (QC tested).