Learn More
ACROBiosystems Biotinylated Human TNFSF11 / RANKL / CD254 Protein, His,Avitag™, active trimer (MALS verified)
Catalog No. 502019617
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human TNFSF11, His,Avitag, active trimer (RAL-H82Q5) is expressed from human 293 cells (HEK293). It contains AA Gly 64 - Asp 245 (Accession # AAC51762.1) trimer Design.;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 24.1 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Measured by its binding ability in a functional ELISA. Immobilized Human RANK, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. RAK-H5251) at 1 μg/mL (100 μL/well) can bind Biotinylated Human TNFSF11, His,Avitag, active trimer (Cat. No. RAL-H82Q5) with a linear range of 0.1-3 ng/mL (QC tested).;Immobilized Human Osteoprotegerin, Fc Tag (Cat. No. TNB-H5259) at 1 μg/mL (100 μL/well) can bind Biotinylated Human TNFSF11, His,Avitag, active trimer (Cat. No. RAL-H82Q5) with a linear range of 0.1-3 ng/mL (Routinely tested).
Catalog No. 50-201-9617 Supplier ACROBiosystems Supplier No. RALH82Q525UG
Recombinant Protein;25UG;Biotinylated Human TNFSF11, His,Avitag, active trimer (RAL-H82Q5) is expressed from human 293 cells (HEK293). It contains AA Gly 64 - Asp 245 (Accession # AAC51762.1) trimer Design.;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 24.1 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Measured by its binding ability in a functional ELISA. Immobilized Human RANK, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. RAK-H5251) at 1 μg/mL (100 μL/well) can bind Biotinylated Human TNFSF11, His,Avitag, active trimer (Cat. No. RAL-H82Q5) with a linear range of 0.1-3 ng/mL (QC tested).;Immobilized Human Osteoprotegerin, Fc Tag (Cat. No. TNB-H5259) at 1 μg/mL (100 μL/well) can bind Biotinylated Human TNFSF11, His,Avitag, active trimer (Cat. No. RAL-H82Q5) with a linear range of 0.1-3 ng/mL (Routinely tested).