Learn More
ACROBiosystems Biotinylated Human CD59 Protein, His,Avitag™
Supplier:  ACROBiosystems CD9H82E325UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human CD59, His,Avitag (CD9-H82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Asn 102 (Accession # P13987-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 12.5 kDa. The protein migrates as 15-20 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9629