Learn More
ACROBiosystems Biotinylated Human GUCY2C / Guanylyl cyclase C Protein, Fc,Avitag™
Catalog No. 502019647
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human GUCY2C, Fc,Avitag (GUC-H82F8) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Gln 430 (Accession # P25092-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 74.1 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Monoclonal Anti-Human GUCY2C Antibody, Human IgG1 at 1 μg/mL (100 μL/well) can bind Biotinylated Human GUCY2C, Fc,Avitag (Cat. No. GUC-H82F8) with a linear range of 0.8-25 ng/mL (QC tested).;Immobilized Biotinylated Human GUCY2C, Fc,Avitag (Cat. No. GUC-H82F8) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human GUCY2C Antibody, Human IgG1 with a linear range of 0.2-3 ng/mL (Routinely tested).
Catalog No. 50-201-9647 Supplier ACROBiosystems Supplier No. GUCH82F825UG
Recombinant Protein;25UG;Biotinylated Human GUCY2C, Fc,Avitag (GUC-H82F8) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Gln 430 (Accession # P25092-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 74.1 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Monoclonal Anti-Human GUCY2C Antibody, Human IgG1 at 1 μg/mL (100 μL/well) can bind Biotinylated Human GUCY2C, Fc,Avitag (Cat. No. GUC-H82F8) with a linear range of 0.8-25 ng/mL (QC tested).;Immobilized Biotinylated Human GUCY2C, Fc,Avitag (Cat. No. GUC-H82F8) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human GUCY2C Antibody, Human IgG1 with a linear range of 0.2-3 ng/mL (Routinely tested).