Learn More
ACROBiosystems Biotinylated Mouse CD8 alpha / CD8A Protein, His,Avitag™
Supplier:  ACROBiosystems CDAM82E825UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Mouse CD8 alpha, His,Avitag (CDA-M82E8) is expressed from human 293 cells (HEK293). It contains AA Lys 28 - Tyr 196 (Accession # P01731-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 22.5 kDa. The protein migrates as 34-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9723