Learn More
ACROBiosystems Biotinylated Human CD36 / SR-B3 Protein, His,Avitag™
Supplier: ACROBiosystems CD6H82E925UG
Recombinant Protein;25UG;Biotinylated Human CD36, His,Avitag (CD6-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Asn 439 (Accession # NP_001001547.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 50.2 kDa.