Learn More
ACROBiosystems Biotinylated Human PVRIG Protein, His,Avitag™
Supplier:  ACROBiosystems PVGH82E325UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human PVRIG, His,Avitag (PVG-H82E3) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession # Q6DKI7-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 17.2 kDa. The protein migrates as 25-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human PVRIG, His,Avitag (Cat. No. PVG-H82E3) at 1 μg/mL (100 μL/well) on Streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Human Nectin-2, Fc Tag (Cat. No. PV2-H5253) with a linear range of 0.1-5 ng/mL (QC tested).
Catalog No. 50-201-9797