Learn More
ACROBiosystems Biotinylated Human FGL1 (64-312) Protein, Avitag™,Fc Tag (high sensitivity)
Supplier:  ACROBiosystems FG1H82F625UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human FGL1 (64-312), Avitag,Fc Tag (FG1-H82F6) is expressed from human 293 cells (HEK293). It contains AA Asp 64 - Ile 312 (Accession # Q08830-1).;This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 116.3 kDa. The protein migrates as 110-120 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human LAG-3, Mouse IgG2a Fc Tag (Cat. No. LA3-H52Aa) at 5 μg/mL (100 μL/well) can bind Biotinylated Human FGL1 (64-312), Avitag,Fc Tag (Cat. No. FG1-H82F6) with a linear range of 0.039-0.313 μg/mL (QC tested).
Catalog No. 50-201-9813