Learn More
ACROBiosystems Biotinylated Human VEGF R2 / KDR Protein, Avitag™,His Tag
Supplier:  ACROBiosystems KDRH82E525UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human VEGF R2, Avitag,His Tag (KDR-H82E5) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Accession # AAI31823).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 85.9 kDa. The protein migrates as 120-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human VEGF165, Tag Free (HPLC-verified) (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Biotinylated Human VEGF R2, Avitag,His Tag (Cat. No. KDR-H82E5) with a linear range of 10-156 ng/mL (QC tested).;Immobilized ActiveMax Human VEGF165, Tag Free (HPLC-verified) (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind pre-mixed increasing concentrations of Bevacizumab and 0.5 μg/mL (100 μL/well) Biotinylated Human VEGF R2, Avitag,His Tag (Cat. No. KDR-H82E5) with a half maximal inhibitory concentration (IC50) of 0.70 μg/mL (Routinely tested).
Catalog No. 50-201-9846