Learn More
ACROBiosystems Biotinylated Human Angiopoietin-like 3 / ANGPTL3 Protein, His,Avitag™
Supplier: ACROBiosystems AN3H82E725UG
Recombinant Protein;25UG;Biotinylated Human Angiopoietin-like 3, His,Avitag (AN3-H82E7) is expressed from human 293 cells (HEK293). It contains AA Ser 17 - Pro 220 (Accession # Q9Y5C1-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 27.4 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human Angiopoietin-like 3, His,Avitag (Cat. No. AN3-H82E7) at 1 μg/mL (100 μL/well) on Streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Monoclonal Anti-Human ANGPTL3 Antibody, Human IgG1 with a linear range of 0.1-2 ng/mL (QC tested).