Learn More
ACROBiosystems FITC-Labeled Human FAP Protein, His Tag
Catalog No. 502019480
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human FAP, Fc Tag (FAP-HF243) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Asp 760 (Accession # Q12884-1). It is the FITC labeled form of Human FAP, His Tag (FAP-H5244).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 86.9 kDa.
Catalog No. 50-201-9480 Supplier ACROBiosystems Supplier No. FAPHF24325UG
Recombinant Protein;25UG;FITC-Labeled Human FAP, Fc Tag (FAP-HF243) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Asp 760 (Accession # Q12884-1). It is the FITC labeled form of Human FAP, His Tag (FAP-H5244).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 86.9 kDa.