Learn More
ACROBiosystems FITC-Labeled Human GUCY2C / Guanylyl cyclase C Protein, His Tag
Catalog No. 502019749
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human GUCY2C, His Tag (GUC-HF2H8) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Gln 430 (Accession # P25092-1). It is the FITC labeled form of Human GUCY2C, His Tag (GUC-H52H5).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.9 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Monoclonal Anti-Human GUCY2C Antibody, Human IgG1 at 5 μg/mL (100 μL/well) can bind FITC-Labeled Human GUCY2C, His Tag (Cat. No. GUC-HF2H8) with a linear range of 0.156-0.625 μg/mL (QC tested).
Catalog No. 50-201-9749 Supplier ACROBiosystems Supplier No. GUCHF2H825UG
Recombinant Protein;25UG;FITC-Labeled Human GUCY2C, His Tag (GUC-HF2H8) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Gln 430 (Accession # P25092-1). It is the FITC labeled form of Human GUCY2C, His Tag (GUC-H52H5).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.9 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Monoclonal Anti-Human GUCY2C Antibody, Human IgG1 at 5 μg/mL (100 μL/well) can bind FITC-Labeled Human GUCY2C, His Tag (Cat. No. GUC-HF2H8) with a linear range of 0.156-0.625 μg/mL (QC tested).