Learn More
ACROBiosystems FITC-Labeled Human ROR1 Protein, Fc Tag
Catalog No. 502019508
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human ROR1, Fc Tag (RO1-HF253) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q01973-1). It is the FITC labeled form of Human ROR1, Fc Tag (RO1-H5250).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 68.6 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Human ROR1 Mab at 1 μg/mL (100 μL/well) can bind FITC-Labeled Human ROR1, Fc Tag (Cat. No. RO1-HF253) with a linear range of 39-313 ng/mL (QC tested).
Catalog No. 50-201-9508 Supplier ACROBiosystems Supplier No. RO1HF25325UG
Recombinant Protein;25UG;FITC-Labeled Human ROR1, Fc Tag (RO1-HF253) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q01973-1). It is the FITC labeled form of Human ROR1, Fc Tag (RO1-H5250).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 68.6 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Human ROR1 Mab at 1 μg/mL (100 μL/well) can bind FITC-Labeled Human ROR1, Fc Tag (Cat. No. RO1-HF253) with a linear range of 39-313 ng/mL (QC tested).